Anti-Tropomyosin Mouse Monoclonal Antibody [clone: TM-36]

Supplier: Boster
BSBTMA1096EA 550 EUR
BSBTMA1096
Anti-Tropomyosin Mouse Monoclonal Antibody [clone: TM-36]
Antibodies
Mouse IgG monoclonal antibody for Tropomyosin (36 kD) detection. Tested with WB, IHC-F in Human;rabbit;chicken. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Tropomyosin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rabbit, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR