Anti-Fibronectin Mouse Monoclonal Antibody [clone: FN3E2]

Supplier: Boster
BSBTMA1116EA 550 EUR
BSBTMA1116
Anti-Fibronectin Mouse Monoclonal Antibody [clone: FN3E2]
Antibodies
Type: Primary
Antigen: Fibronectin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chicken, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR