Imprimer

Anti-Annexin A1 Rabbit Polyclonal Antibody

Fournisseur: Boster
BSBTPA1006EA 550 EUR
BSBTPA1006
Anti-Annexin A1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Annexin A1 (ANXA1) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Annexin A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR