Anti-NMDAR2A Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPA1058-1EA 0 EUR
BSBTPA1058-1
Anti-NMDAR2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NMDAR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR