Anti-CX3CL1 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPA1401EA 550 EUR
BSBTPA1401
Anti-CX3CL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Fractalkine(CX3CL1) detection. Tested with WB, IHC-P, IHC-F in Mouse; Rat.

  • Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CX3CL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR