Anti-Progesterone Receptor Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPA1413EA 550 EUR
BSBTPA1413
Anti-Progesterone Receptor Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Progesterone receptor(PGR) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Progesterone Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR