Anti-Galectin 1 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPA1422EA 550 EUR
BSBTPA1422
Anti-Galectin 1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Galectin 1(LGALS1) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Galectin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR