Anti-IL2 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPA1466EA 550 EUR
BSBTPA1466
Anti-IL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Interleukin-2(IL2) detection. Tested with WB in Mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR