Anti-Fos B Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPA1478EA 550 EUR
BSBTPA1478
Anti-Fos B Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Protein fosB(FOSB) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Fos B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR