Anti-Ribonuclease A Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPA1507EA 550 EUR
BSBTPA1507
Anti-Ribonuclease A Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Ribonuclease pancreatic(RNASE1) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Ribonuclease A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR