Imprimer

Anti-Ig Sheep Polyclonal Antibody (Biotin)

Fournisseur: Cytiva
RPN1001-2MLEA 621 EUR
RPN1001-2ML
Anti-Ig Sheep Polyclonal Antibody (Biotin)
Anticorps
The antibodies are supplied with dilution guidelines for relevant applications.

  • Soluble in cold water and hot water

The antibodies are supplied with dilution guidelines for relevant applications and are stored at 2°C to 8°C.

Type: Secondary
Antigen: Mouse Ig
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR