Imprimer

Anti-LAG3 Mouse Monoclonal Antibody [clone: 17B4]

Fournisseur: United States Biological
USBI044991EA 527 EUR
USBI044991
Anti-LAG3 Mouse Monoclonal Antibody [clone: 17B4]
Anticorps
Recommended dilution: Immunochemistry: 1:150; Immunoprecipitation: 10 ?g/ml; Western Blot: 5 ?g/ml

Type: Primary
Antigen: LAG3 (lymphocyte-activation gene 3)
Clonality: Monoclonal
Clone: 17B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR