Imprimer

Anti-FBXW7 Mouse Monoclonal Antibody [clone: 2G7]

Fournisseur: OriGene
ORIGTA802834EA 603 EUR
ORIGTA802834
Anti-FBXW7 Mouse Monoclonal Antibody [clone: 2G7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: FBXW7
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR