Anti-FICD Mouse Monoclonal Antibody [clone: 2A11]

Supplier: OriGene
ORIGTA802841EA 603 EUR
ORIGTA802841
Anti-FICD Mouse Monoclonal Antibody [clone: 2A11]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: FICD
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR