Anti-CD68 Mouse Monoclonal Antibody [clone: 2H10]

Supplier: OriGene
ORIGTA802851EA 603 EUR
ORIGTA802851
Anti-CD68 Mouse Monoclonal Antibody [clone: 2H10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 2H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR