Anti-HADH Mouse Monoclonal Antibody [clone: 1E1]

Supplier: OriGene
ORIGTA802888EA 603 EUR
ORIGTA802888
Anti-HADH Mouse Monoclonal Antibody [clone: 1E1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HADH
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR