Anti-HADH Mouse Monoclonal Antibody [clone: 1D4]

Supplier: OriGene
ORIGTA802894EA 603 EUR
ORIGTA802894
Anti-HADH Mouse Monoclonal Antibody [clone: 1D4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: HADH
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR