Anti-KRT24 Mouse Monoclonal Antibody [clone: 4C3]

Supplier: OriGene
ORIGTA802895EA 603 EUR
ORIGTA802895
Anti-KRT24 Mouse Monoclonal Antibody [clone: 4C3]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: KRT24
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR