Anti-NAB1 Mouse Monoclonal Antibody [clone: 4G10]

Supplier: OriGene
ORIGTA802913EA 603 EUR
ORIGTA802913
Anti-NAB1 Mouse Monoclonal Antibody [clone: 4G10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NAB1
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR