Anti-NTRK3 Mouse Monoclonal Antibody [clone: 24B3]

Supplier: OriGene
ORIGCF500536EA 749 EUR
ORIGCF500536
Anti-NTRK3 Mouse Monoclonal Antibody [clone: 24B3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: NTRK3
Clonality: Monoclonal
Clone: 24B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR