Anti-UBXN2B Mouse Monoclonal Antibody [clone: 1G9]

Supplier: OriGene
ORIGCF502489EA 770 EUR
ORIGCF502489
Anti-UBXN2B Mouse Monoclonal Antibody [clone: 1G9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: UBXN2B
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR