Anti-VASP Mouse Monoclonal Antibody [clone: 1F3]

Supplier: OriGene
ORIGCF502491EA 770 EUR
ORIGCF502491
Anti-VASP Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150

Type: Primary
Antigen: VASP
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR