Anti-S100A12 Mouse Monoclonal Antibody [clone: 1D1]

Supplier: OriGene
ORIGCF502495EA 749 EUR
ORIGCF502495
Anti-S100A12 Mouse Monoclonal Antibody [clone: 1D1]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: S100A12
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR