Anti-ATP5B Mouse Monoclonal Antibody [clone: 1F7]

Supplier: OriGene
ORIGCF500832EA 749 EUR
ORIGCF500832
Anti-ATP5B Mouse Monoclonal Antibody [clone: 1F7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: ATP5B
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR