Anti-GRIPAP1 Mouse Monoclonal Antibody [clone: 1F4]

Supplier: OriGene
ORIGCF500854EA 770 EUR
ORIGCF500854
Anti-GRIPAP1 Mouse Monoclonal Antibody [clone: 1F4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GRIPAP1
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR