Anti-LOX Mouse Monoclonal Antibody [clone: 4E3]

Supplier: OriGene
ORIGCF500865EA 770 EUR
ORIGCF500865
Anti-LOX Mouse Monoclonal Antibody [clone: 4E3]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LOX
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR