Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 2A5]

Supplier: OriGene
ORIGCF501795EA 749 EUR
ORIGCF501795
Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 2A5]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EPM2AIP1
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR