Anti-GSTT2 Mouse Monoclonal Antibody [clone: 7A12]

Supplier: OriGene
ORIGCF501858EA 770 EUR
ORIGCF501858
Anti-GSTT2 Mouse Monoclonal Antibody [clone: 7A12]
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:2000; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 7A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR