Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 7G6]

Supplier: OriGene
ORIGCF501867EA 749 EUR
ORIGCF501867
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 7G6]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH1L1
Clonality: Monoclonal
Clone: 7G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR