Anti-RTN4IP1 Mouse Monoclonal Antibody [clone: 3B2]

Supplier: OriGene
ORIGCF502544EA 749 EUR
ORIGCF502544
Anti-RTN4IP1 Mouse Monoclonal Antibody [clone: 3B2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: RTN4IP1
Clonality: Monoclonal
Clone: 3B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR