Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: 3B5]

Supplier: OriGene
ORIGCF500883EA 749 EUR
ORIGCF500883
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: 3B5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: HSP90AA1
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR