Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 1D12]

Supplier: OriGene
ORIGCF501502EA 749 EUR
ORIGCF501502
Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 1D12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MOBKL2B
Clonality: Monoclonal
Clone: 1D12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR