Anti-BTN1A1 Mouse Monoclonal Antibody [clone: 3A12]

Supplier: OriGene
ORIGCF501530EA 749 EUR
ORIGCF501530
Anti-BTN1A1 Mouse Monoclonal Antibody [clone: 3A12]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BTN1A1
Clonality: Monoclonal
Clone: 3A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR