Anti-BTN1A1 Mouse Monoclonal Antibody [clone: 5E6]

Supplier: OriGene
ORIGCF501536EA 749 EUR
ORIGCF501536
Anti-BTN1A1 Mouse Monoclonal Antibody [clone: 5E6]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: BTN1A1
Clonality: Monoclonal
Clone: 5E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR