Anti-RNH1 Mouse Monoclonal Antibody [clone: 4G4]

Supplier: OriGene
ORIGCF501874EA 749 EUR
ORIGCF501874
Anti-RNH1 Mouse Monoclonal Antibody [clone: 4G4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNH1
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR