Anti-ZIM2 Mouse Monoclonal Antibody [clone: 5E12]

Supplier: OriGene
ORIGCF501906EA 770 EUR
ORIGCF501906
Anti-ZIM2 Mouse Monoclonal Antibody [clone: 5E12]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ZIM2
Clonality: Monoclonal
Clone: 5E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR