Anti-HDHD1A Mouse Monoclonal Antibody [clone: 3E5]

Supplier: OriGene
ORIGCF502557EA 749 EUR
ORIGCF502557
Anti-HDHD1A Mouse Monoclonal Antibody [clone: 3E5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HDHD1A
Clonality: Monoclonal
Clone: 3E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR