Anti-MEF2C Mouse Monoclonal Antibody [clone: 2F2]

Supplier: OriGene
ORIGCF502552EA 749 EUR
ORIGCF502552
Anti-MEF2C Mouse Monoclonal Antibody [clone: 2F2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: MEF2C
Clonality: Monoclonal
Clone: 2F2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR