Anti-ANKRD53 Mouse Monoclonal Antibody [clone: 2A8]

Supplier: OriGene
ORIGCF502599EA 749 EUR
ORIGCF502599
Anti-ANKRD53 Mouse Monoclonal Antibody [clone: 2A8]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ANKRD53
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR