Anti-CD36 Mouse Monoclonal Antibody [clone: 4H7]

Supplier: OriGene
ORIGCF500935EA 749 EUR
ORIGCF500935
Anti-CD36 Mouse Monoclonal Antibody [clone: 4H7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 4H7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR