Anti-SRR Mouse Monoclonal Antibody [clone: 1D7]

Supplier: OriGene
ORIGCF500942EA 749 EUR
ORIGCF500942
Anti-SRR Mouse Monoclonal Antibody [clone: 1D7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: SRR
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR