Anti-NPR3 Mouse Monoclonal Antibody [clone: 2C2]

Supplier: OriGene
ORIGCF500955EA 749 EUR
ORIGCF500955
Anti-NPR3 Mouse Monoclonal Antibody [clone: 2C2]
Antibodies
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: NPR3
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR