Anti-KIAA1609 Mouse Monoclonal Antibody [clone: 5D9]

Supplier: OriGene
ORIGCF500962EA 770 EUR
ORIGCF500962
Anti-KIAA1609 Mouse Monoclonal Antibody [clone: 5D9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: KIAA1609
Clonality: Monoclonal
Clone: 5D9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR