Anti-FAM84B Mouse Monoclonal Antibody [clone: 5A9]

Supplier: OriGene
ORIGCF501918EA 749 EUR
ORIGCF501918
Anti-FAM84B Mouse Monoclonal Antibody [clone: 5A9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FAM84B
Clonality: Monoclonal
Clone: 5A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR