Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 6H9]

Supplier: OriGene
ORIGCF501939EA 749 EUR
ORIGCF501939
Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 6H9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: OSBPL11
Clonality: Monoclonal
Clone: 6H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR