Anti-OTUB2 Mouse Monoclonal Antibody [clone: 11A5]

Supplier: OriGene
ORIGCF501941EA 749 EUR
ORIGCF501941
Anti-OTUB2 Mouse Monoclonal Antibody [clone: 11A5]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: OTUB2
Clonality: Monoclonal
Clone: 11A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR