Anti-SAMHD1 Mouse Monoclonal Antibody [clone: 1F9]

Supplier: OriGene
ORIGCF501955EA 749 EUR
ORIGCF501955
Anti-SAMHD1 Mouse Monoclonal Antibody [clone: 1F9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SAMHD1
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR