Anti-ANKRD53 Mouse Monoclonal Antibody [clone: 1E1]

Supplier: OriGene
ORIGCF502601EA 749 EUR
ORIGCF502601
Anti-ANKRD53 Mouse Monoclonal Antibody [clone: 1E1]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ANKRD53
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR