Anti-PON1 Mouse Monoclonal Antibody [clone: 2A9]

Supplier: OriGene
ORIGCF502612EA 749 EUR
ORIGCF502612
Anti-PON1 Mouse Monoclonal Antibody [clone: 2A9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PON1
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR