Anti-TPMT Mouse Monoclonal Antibody [clone: 5B4]

Supplier: OriGene
ORIGCF500965EA 749 EUR
ORIGCF500965
Anti-TPMT Mouse Monoclonal Antibody [clone: 5B4]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: TPMT
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR